Tisk…

2,5-Dichlorobenzothiazole 97%

Dodavatel: Apollo Scientific

OR480498-5G
APOSOR480498-5GEA 3540 CZK
APOSOR480498-5G
2,5-Dichlorobenzothiazole 97%
2,5-Dichlorobenzothiazole
Vzorec: C₇H₃Cl₂NS
MW: 204.0764 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044024
CAS číslo: 2941-48-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR