Tisk…

2-Amino-6-isopropylpyrimidin-4-ol 95%

Dodavatel: Apollo Scientific

OR480221-1G OR480221-5G
APOSOR480221-1GEA 8200 CZK
APOSOR480221-1G APOSOR480221-5G
2-Amino-6-isopropylpyrimidin-4-ol 95%
2-Amino-6-isopropylpyrimidin-4-ol
Vzorec: C₇H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD02091393
CAS číslo: 73576-32-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR