Tisk…

4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%

Dodavatel: Apollo Scientific

OR480203-1G OR480203-5G
APOSOR480203-1GEA 5000 CZK
APOSOR480203-1G APOSOR480203-5G
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester
Vzorec: C₈H₇NO₂S₂
MW: 213.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173757
CAS číslo: 175202-48-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR