Tisk…

Ethyl 5-amino-4-chloro-1-(cyclopropylmethyl)pyrazole-3-carboxylate

Dodavatel: Apollo Scientific

OR480033-1G
APOSOR480033-1GEA 32440 CZK
APOSOR480033-1G
Ethyl 5-amino-4-chloro-1-(cyclopropylmethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(cyclopropylmethyl)pyrazole-3-carboxylate
Vzorec: C₁₀H₁₄ClN₃O₂
MW: 243,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135199
CAS číslo: 1427013-88-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR