Tisk…

5-(1H-Pyrrol-1-yl)nicotinic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(1H-Pyrrol-1-yl)pyridine-3-carboxylic acid

OR4766-250MG
APOSOR4766-250MGEA 2980 CZK
APOSOR4766-250MG
5-(1H-Pyrrol-1-yl)nicotinic acid 95%
5-(1H-Pyrrol-1-yl)nicotinic acid
Vzorec: C₁₀H₈N₂O₂
MW: 188.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971938
CAS číslo: 690632-31-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR