Tisk…

N-(2-Chloropyridin-4-yl)acetamide 98%

Dodavatel: Apollo Scientific

OR4744-1G
APOSOR4744-1GEA 1620 CZK
APOSOR4744-1G
N-(2-Chloropyridin-4-yl)acetamide 98%
N-(2-Chloropyridin-4-yl)acetamide
Vzorec: C₇H₇ClN₂O
MW: 170,6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234294
CAS číslo: 13602-82-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR