Tisk…

Methyl 2-(6-bromo-3-pyridyl)acetate 95%

Dodavatel: Apollo Scientific

OR471726-1G OR471726-250MG
APOSOR471726-1GEA 14620 CZK
APOSOR471726-1G APOSOR471726-250MG
Methyl 2-(6-bromo-3-pyridyl)acetate 95%
Methyl 2-(6-bromo-3-pyridyl)acetate
Vzorec: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD18256344
CAS číslo: 1256824-78-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR