Tisk…

2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR471723-100MG OR471723-1G OR471723-250MG
APOSOR471723-100MGEA 19080 CZK
APOSOR471723-100MG APOSOR471723-1G APOSOR471723-250MG
2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde 95%
2-(Cyclopropylmethoxy)pyrimidine-5-carbaldehyde
Vzorec: C₉H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26843559
CAS číslo: 1192569-90-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR