Tisk…

2-Amino-3-cyanopyridine-5-boronic acid pinacol ester 95%

Dodavatel: Apollo Scientific

OR471722-1G OR471722-250MG
APOSOR471722-1GEA 7440 CZK
APOSOR471722-1G APOSOR471722-250MG
2-Amino-3-cyanopyridine-5-boronic acid pinacol ester 95%
2-Amino-3-cyanopyridine-5-boronic acid pinacol ester
Vzorec: C₁₂H₁₆BN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD16995200
CAS číslo: 1246372-66-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR