Tisk…

5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 95%

Dodavatel: Apollo Scientific
Varování

OR471704-1G OR471704-250MG
APOSOR471704-1GEA 1780 CZK
APOSOR471704-1G APOSOR471704-250MG
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 95%
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine
Vzorec: C₁₁H₁₆IN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27978466
CAS číslo: 661487-17-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR