Tisk…

Methyl 6-bromoindole-3-carboxylate 97%

Dodavatel: Apollo Scientific

OR471662-5G
APOSOR471662-5GEA 4240 CZK
APOSOR471662-5G
Methyl 6-bromoindole-3-carboxylate 97%
Methyl 6-bromoindole-3-carboxylate
Vzorec: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD09836005
CAS číslo: 868656-97-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR