Tisk…

L-Threonine benzyl ester hydrochloride 98%

Dodavatel: Apollo Scientific

Synonyms: (2S,3R)-Benzyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OBzl.HCl

OR471632-25G
APOSOR471632-25GEA 3580 CZK
APOSOR471632-25G
L-Threonine benzyl ester hydrochloride 98%
L-Threonine benzyl ester hydrochloride
Vzorec: C₁₁H₁₅NO₃
MW: 245.70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038977
CAS číslo: 33645-24-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR