Tisk…

1,4-Dibenzyloxy-2-nitrobenzene

Dodavatel: Apollo Scientific

OR471569-25G
APOSOR471569-25GEA 9030 CZK
APOSOR471569-25G
1,4-Dibenzyloxy-2-nitrobenzene
1,4-Dibenzyloxy-2-nitrobenzene
Vzorec: C₂₀H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00082778
CAS číslo: 51792-85-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR