Tisk…

2-(Methylthio)-4,5-pyrimidinedicarboxylic acid

Dodavatel: Apollo Scientific

OR471456-250MG OR471456-1G OR471456-5G
APOSOR471456-250MGEA 5820 CZK
APOSOR471456-250MG APOSOR471456-1G APOSOR471456-5G
2-(Methylthio)-4,5-pyrimidinedicarboxylic acid
2-(Methylthio)-4,5-pyrimidinedicarboxylic acid
Vzorec: C₇H₆N₂O₄S
MW: 214.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195439
CAS číslo: 149771-16-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR