Tisk…

1-(Methylamino)-3-(4-methylpiperazin-1-yl)-2-propanol

Dodavatel: Apollo Scientific

OR471418-1G
APOSOR471418-1GEA 13920 CZK
APOSOR471418-1G
1-(Methylamino)-3-(4-methylpiperazin-1-yl)-2-propanol
1-(Methylamino)-3-(4-methylpiperazin-1-yl)-2-propanol
Vzorec: C₉H₂₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD00776867
CAS číslo: 462066-61-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR