Tisk…

N-(2-Chloro-4-methyl-3-pyridyl)acetamide

Dodavatel: Apollo Scientific

OR471339-1G OR471339-5G
APOSOR471339-1GEA 6980 CZK
APOSOR471339-1G APOSOR471339-5G
N-(2-Chloro-4-methyl-3-pyridyl)acetamide
N-(2-Chloro-4-methyl-3-pyridyl)acetamide
Vzorec: C₈H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD21244018
CAS číslo: 76006-00-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR