Tisk…

Methyl methyl-4-nitro-1H-imidazole-2-carboxylate

Dodavatel: Apollo Scientific

Synonyms: Methyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

OR471118-5G
APOSOR471118-5GEA 11250 CZK
APOSOR471118-5G
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate
Vzorec: C₆H₇N₃O₄
MW: 185.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275078
CAS číslo: 169770-25-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR