Tisk…

2-(2-Iodo-4-nitrophenyl)acetic acid

Dodavatel: Apollo Scientific

OR471044-1G OR471044-250MG
APOSOR471044-1GEA 26090 CZK
APOSOR471044-1G APOSOR471044-250MG
2-(2-Iodo-4-nitrophenyl)acetic acid
2-(2-Iodo-4-nitrophenyl)acetic acid
Vzorec: C₈H₆INO₄
Storage Temperature: Ambient
MDL Number: MFCD18398041
CAS číslo: 66949-41-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR