Tisk…

Methyl 4-[Imino(methoxy)methyl]benzoate hydrochloride

Dodavatel: Apollo Scientific

OR470980-1G
APOSOR470980-1GEA 5390 CZK
APOSOR470980-1G
Methyl 4-[Imino(methoxy)methyl]benzoate hydrochloride
Methyl 4-[Imino(methoxy)methyl]benzoate hydrochloride
Vzorec: C₁₀H₁₁NO₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD10697945
CAS číslo: 63617-98-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR