Tisk…

Ethyl 7-Bromoisoquinoline-1-carboxylate

Dodavatel: Apollo Scientific

OR470935-1G
APOSOR470935-1GEA 32770 CZK
APOSOR470935-1G
Ethyl 7-Bromoisoquinoline-1-carboxylate
Ethyl 7-Bromoisoquinoline-1-carboxylate
Vzorec: C₁₂H₁₀BrNO₂
MW: 280,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986818
CAS číslo: 1823278-31-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR