Tisk…

2-[2-(Methylsulphonyl)phenyl]ethylamine hydrochloride

Dodavatel: Apollo Scientific

OR470914-5G
APOSOR470914-5GEA 28700 CZK
APOSOR470914-5G
2-[2-(Methylsulphonyl)phenyl]ethylamine hydrochloride
2-[2-(Methylsulphonyl)phenyl]ethylamine hydrochloride
Vzorec: C₉H₁₃NO₂S·ClH
Storage Temperature: Ambient
MDL Number: MFCD27986816
CAS číslo: 2006277-03-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR