Tisk…

4-Chlorohexanoyl chloride

Dodavatel: Apollo Scientific

OR470809-5G OR470809-1G
APOSOR470809-5GEA 22920 CZK
APOSOR470809-5G APOSOR470809-1G
4-Chlorohexanoyl chloride
4-Chlorohexanoyl chloride
Vzorec: C₆H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD20622929
CAS číslo: 99585-00-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR