Tisk…

3-(2-Pyridyl)propiolic acid

Dodavatel: Apollo Scientific

OR470616-5G OR470616-1G
APOSOR470616-5GEA 28700 CZK
APOSOR470616-5G APOSOR470616-1G
3-(2-Pyridyl)propiolic acid
3-(2-Pyridyl)propiolic acid
Vzorec: C₈H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD16619698
CAS číslo: 858678-71-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR