Tisk…

Ethyl 3-(4-Pyridyl)propanoate

Dodavatel: Apollo Scientific

OR470608-5G
APOSOR470608-5GEA 20300 CZK
APOSOR470608-5G
Ethyl 3-(4-Pyridyl)propanoate
Ethyl 3-(4-Pyridyl)propanoate
Vzorec: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD11045822
CAS číslo: 52809-19-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR