Tisk…

N-(3-Indolinylmethyl)benzylamine

Dodavatel: Apollo Scientific

OR470604-1G
APOSOR470604-1GEA 17390 CZK
APOSOR470604-1G
N-(3-Indolinylmethyl)benzylamine
N-(3-Indolinylmethyl)benzylamine
Vzorec: C₁₆H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD23381108
CAS číslo: 1427475-17-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR