Tisk…

2-Amino-2-(4-biphenylyl)acetic acid hydrochloride

Dodavatel: Apollo Scientific

OR470371-5G
APOSOR470371-5GEA 43640 CZK
APOSOR470371-5G
2-Amino-2-(4-biphenylyl)acetic acid hydrochloride
2-Amino-2-(4-biphenylyl)acetic acid hydrochloride
Vzorec: C₁₄H₁₃NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD06202322
CAS číslo: 885498-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR