Tisk…

5-Hydroxy-2-(methylsulphonamido)benzoic acid

Dodavatel: Apollo Scientific

OR470319-1G OR470319-5G
APOSOR470319-1GEA 7840 CZK
APOSOR470319-1G APOSOR470319-5G
5-Hydroxy-2-(methylsulphonamido)benzoic acid
5-Hydroxy-2-(methylsulphonamido)benzoic acid
Vzorec: C₈H₉NO₅S
Storage Temperature: Ambient
MDL Number: MFCD14660388
CAS číslo: 1243391-74-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR