Tisk…

Ethyl 4-(2-Bromophenyl)thiazole-2-carboxylate

Dodavatel: Apollo Scientific

OR470185-5G
APOSOR470185-5GEA 22920 CZK
APOSOR470185-5G
Ethyl 4-(2-Bromophenyl)thiazole-2-carboxylate
Ethyl 4-(2-Bromophenyl)thiazole-2-carboxylate
Vzorec: C₁₂H₁₀BrNO₂S
MW: 312,182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846637
CAS číslo: 1261269-07-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR