Tisk…

2,6-Bis[1-[(2,6-dimethylphenyl)imino]ethyl]pyridine

Dodavatel: Apollo Scientific

OR470167-5G
APOSOR470167-5GEA 43640 CZK
APOSOR470167-5G
2,6-Bis[1-[(2,6-dimethylphenyl)imino]ethyl]pyridine
2,6-Bis[1-[(2,6-dimethylphenyl)imino]ethyl]pyridine
Vzorec: C₂₅H₂₇N₃
Storage Temperature: Ambient
MDL Number: MFCD18375270
CAS číslo: 204203-16-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR