Tisk…

2-Chloro-4-morpholinobenzenecarboxylic acid

Dodavatel: Apollo Scientific

OR470071-1G OR470071-5G
APOSOR470071-1GEA 0 CZK
APOSOR470071-1G APOSOR470071-5G
2-Chloro-4-morpholinobenzenecarboxylic acid
2-Chloro-4-morpholinobenzenecarboxylic acid
Vzorec: C₁₁H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD03425732
CAS číslo: 175153-55-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR