Tisk…

6-(Cbz-amino)-3-pyridazinecarboxylic acid

Dodavatel: Apollo Scientific

OR470060-1G
APOSOR470060-1GEA 10150 CZK
APOSOR470060-1G
6-(Cbz-amino)-3-pyridazinecarboxylic acid
6-(Cbz-amino)-3-pyridazinecarboxylic acid
Vzorec: C₁₃H₁₁N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD13191593
CAS číslo: 1197193-41-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR