Tisk…

3-Phenylisonicotinic acid

Dodavatel: Apollo Scientific

OR46655-1G OR46655-100MG OR46655-250MG OR46655-500MG OR46655-2.5G OR46655-5G
APOSOR46655-1GEA 16770 CZK
APOSOR46655-1G APOSOR46655-100MG APOSOR46655-250MG APOSOR46655-500MG APOSOR46655-2.5G APOSOR46655-5G
3-Phenylisonicotinic acid
3-Phenylisonicotinic acid
Vzorec: C₁₂H₉NO₂
MW: 199.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114253
CAS číslo: 104096-15-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR