Tisk…

2,2'-Dithiodianiline (thulium ionophore I) 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR4664-100G OR4664-500G
APOSOR4664-100GEA 2570 CZK
APOSOR4664-100G APOSOR4664-500G
2,2'-Dithiodianiline (thulium ionophore I) 98%
2,2'-Dithiodianiline (thulium ionophore I)
Vzorec: C₁₂H₁₂N₂S₂
Bod tání: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007703
CAS číslo: 1141-88-4
EINECS: 214-529-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR