Tisk…

3-(tert-Butoxy)benzeneboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(tert-Butoxy)phenyl)boronic acid

OR46618-1G OR46618-250MG OR46618-5G
APOSOR46618-1GEA 7250 CZK
APOSOR46618-1G APOSOR46618-250MG APOSOR46618-5G
3-(tert-Butoxy)benzeneboronic acid
3-(tert-Butoxy)benzeneboronic acid
Vzorec: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06801745
CAS číslo: 312931-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR