Tisk…

4-(Methylsulphonyl)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR46201-5G OR46201-100G OR46201-25G OR46201-1G
APOSOR46201-5GEA 2280 CZK
APOSOR46201-5G APOSOR46201-100G APOSOR46201-25G APOSOR46201-1G
4-(Methylsulphonyl)benzyl bromide 97%
4-(Methylsulphonyl)benzyl bromide
Vzorec: C₈H₉BrO₂S
Bod tání: 82…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185849
CAS číslo: 53606-06-7
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR