Tisk…

5-Bromo-N-cyclopropylpyridin-2-amine 98%

Dodavatel: Apollo Scientific

OR46049-1G OR46049-5G
APOSOR46049-1GEA 3970 CZK
APOSOR46049-1G APOSOR46049-5G
5-Bromo-N-cyclopropylpyridin-2-amine 98%
5-Bromo-N-cyclopropylpyridin-2-amine
Vzorec: C₈H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08706291
CAS číslo: 885266-96-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR