Tisk…

(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95+%

Dodavatel: Apollo Scientific
Varování

OR460002-250MG OR460002-1G OR460002-5G
APOSOR460002-250MGEA 2280 CZK
APOSOR460002-250MG APOSOR460002-1G APOSOR460002-5G
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95+%
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine
Vzorec: C₁₀H₂₂N₂
Storage Temperature: Ambient
MDL Number: MFCD03411208
CAS číslo: 53152-69-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR