Tisk…

tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate 97%

Dodavatel: Apollo Scientific

OR452185-1G
APOSOR452185-1GEA 3110 CZK
APOSOR452185-1G
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate
Vzorec: C₁₁H₁₉BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD16990579
CAS číslo: 112257-12-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR