Tisk…

3-(Methoxymethyl)piperidine hydrochloride 97%

Dodavatel: Apollo Scientific

OR452125-1G OR452125-500MG
APOSOR452125-1GEA 11180 CZK
APOSOR452125-1G APOSOR452125-500MG
3-(Methoxymethyl)piperidine hydrochloride 97%
3-(Methoxymethyl)piperidine hydrochloride
Vzorec: C₇H₁₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD08166754
CAS číslo: 688809-97-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR