Tisk…

L-Alanine cyclobutylmethyl ester hydrochloride 97%

Dodavatel: Apollo Scientific

OR452064-1G OR452064-5G
APOSOR452064-1GEA 9690 CZK
APOSOR452064-1G APOSOR452064-5G
L-Alanine cyclobutylmethyl ester hydrochloride 97%
L-Alanine cyclobutylmethyl ester hydrochloride
Vzorec: C₈H₁₅NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD27952567
CAS číslo: 261909-76-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR