Tisk…

(3,4-Dichlorophenoxy)acetic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 3,4-D, 2-(3,4-Dichlorophenoxy)acetic acid

OR452057-5G
APOSOR452057-5GEA 2030 CZK
APOSOR452057-5G
(3,4-Dichlorophenoxy)acetic acid 97%
(3,4-Dichlorophenoxy)acetic acid
Vzorec: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04
Storage Temperature: Ambient
MDL Number: MFCD00004303
CAS číslo: 588-22-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR