Tisk…

1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%

Dodavatel: Apollo Scientific

OR452055-1G OR452055-5G
APOSOR452055-1GEA 1370 CZK
APOSOR452055-1G APOSOR452055-5G
1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%
1-(3-Bromophenyl)cyclopentanecarboxylic acid
Vzorec: C₁₂H₁₃BrO₂
MW: 269.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00573784
CAS číslo: 143328-23-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR