Tisk…

6-Methyl-L-tryptophan 97%

Dodavatel: Apollo Scientific

OR452052-250MG
APOSOR452052-250MGEA 10580 CZK
APOSOR452052-250MG
6-Methyl-L-tryptophan 97%
6-Methyl-L-tryptophan
Vzorec: C₁₂H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06797708
CAS číslo: 33468-34-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR