Tisk…

N-Fmoc-3-hydroxy-L-phenylalanine 97%

Dodavatel: Apollo Scientific

OR452043-1G OR452043-250MG OR452043-500MG
APOSOR452043-1GEA 18620 CZK
APOSOR452043-1G APOSOR452043-250MG APOSOR452043-500MG
N-Fmoc-3-hydroxy-L-phenylalanine 97%
N-Fmoc-3-hydroxy-L-phenylalanine
Vzorec: C₂₄H₂₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD07371961
CAS číslo: 178432-48-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR