Tisk…

1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone

Dodavatel: Apollo Scientific
Varování

OR451418-1G OR451418-5G
APOSOR451418-1GEA 992 CZK
APOSOR451418-1G APOSOR451418-5G
1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone
1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone
Vzorec: C₆H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD04968842
CAS číslo: 925200-47-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR