Tisk…

4-Boc-2-(bromomethyl)-morpholine

Dodavatel: Apollo Scientific
Varování

OR451230-1G
APOSOR451230-1GEA 8440 CZK
APOSOR451230-1G
4-Boc-2-(bromomethyl)-morpholine
4-Boc-2-(bromomethyl)-morpholine
Vzorec: C₁₀H₁₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD20489326
CAS číslo: 765914-78-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR