Tisk…

3-(Tetrahydropyran-4-yloxy)benzoic acid

Dodavatel: Apollo Scientific

OR451222-1G
APOSOR451222-1GEA 27350 CZK
APOSOR451222-1G
3-(Tetrahydropyran-4-yloxy)benzoic acid
3-(Tetrahydropyran-4-yloxy)benzoic acid
Vzorec: C₁₂H₁₄O₄
MW: 222.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19611449
CAS číslo: 862088-66-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR