Tisk…

1-(3-Bromophenyl)-cyclobutanemethanamine 95%

Dodavatel: Apollo Scientific

OR451073-1G OR451073-250MG
APOSOR451073-1GEA 15480 CZK
APOSOR451073-1G APOSOR451073-250MG
1-(3-Bromophenyl)-cyclobutanemethanamine 95%
1-(3-Bromophenyl)-cyclobutanemethanamine
Vzorec: C₁₁H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD11642693
CAS číslo: 915690-61-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR