Tisk…

Chroman-6-carbaldehyde

Dodavatel: Apollo Scientific
Varování

OR45040-1G OR45040-250MG
APOSOR45040-1GEA 8700 CZK
APOSOR45040-1G APOSOR45040-250MG
Chroman-6-carbaldehyde
Chroman-6-carbaldehyde
Vzorec: C₁₀H₁₀O₂
Bod varu: 98…104 °C/0,6mmHg
Storage Temperature: Ambient
MDL Number: MFCD08146589
CAS číslo: 55745-97-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR