Tisk…

4-(Methylsulphonyl)phenylhydrazine hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

OR4432-1G
APOSOR4432-1GEA 622 CZK
APOSOR4432-1G
4-(Methylsulphonyl)phenylhydrazine hydrochloride 98%
4-(Methylsulphonyl)phenylhydrazine hydrochloride
Vzorec: C₇H₁₁ClN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00216494
CAS číslo: 17852-67-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR